Methyl (2S,4S)-4-(4-chloro-2-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050976-500MG 050976-2.5G
101893-796EA 364.62 USD
101893-796 101893-798
Methyl (2S,4S)-4-(4-chloro-2-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-chloro-2-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₁Cl₂NO₃
MDL Number: MFCD13561346
CAS Number: 1354488-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR