Methyl (2S,4S)-4-(2-chloro-4-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
050974-500MG
050974-2.5G
101893-788EA
364.62
USD
101893-788
101893-790
Methyl (2S,4S)-4-(2-chloro-4-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-chloro-4-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₉Cl₂NO₃ |
MDL Number:
MFCD13561344 CAS Number: 1354487-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...