Methyl (2S,4S)-4-[(3-chloro[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050966-500MG 050966-2.5G
101893-756EA 364.62 USD
101893-756 101893-758
Methyl (2S,4S)-4-[(3-chloro[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(3-chloro[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₈H₁₉Cl₂NO₃
MDL Number: MFCD13561336
CAS Number: 1354488-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR