(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(3-chloro[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050965-500MG 050965-2.5G
101893-752EA 364.62 USD
101893-752 101893-754
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(3-chloro[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(3-chloro[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₂H₂₄ClNO₅
MW: 417.89 g/mol
MDL Number: MFCD13561335
CAS Number: 1354485-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR