Methyl (2S,4S)-4-(2-chloro-4,6-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050964-500MG 050964-2.5G
101893-748EA 364.62 USD
101893-748 101893-750
Methyl (2S,4S)-4-(2-chloro-4,6-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-chloro-4,6-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉Cl₂NO₃
MDL Number: MFCD13561334
CAS Number: 1354488-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR