(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4,6-dimethylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050963-500MG 050963-2.5G
101893-744EA 364.62 USD
101893-744 101893-746
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4,6-dimethylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4,6-dimethylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₄ClNO₅
MDL Number: MFCD13561333
CAS Number: 1354486-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR