Methyl (2S,4S)-4-[(5-bromo[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050960-500MG 050960-2.5G
101893-732EA 364.62 USD
101893-732 101893-734
Methyl (2S,4S)-4-[(5-bromo[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(5-bromo[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₈H₁₉BrClNO₃
MDL Number: MFCD13561330
CAS Number: 1354487-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR