Methyl (2S,4S)-4-[2-bromo-4-(1-methyl-1-phenylethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050958-500MG 050958-2.5G
101893-724EA 364.62 USD
101893-724 101893-726
Methyl (2S,4S)-4-[2-bromo-4-(1-methyl-1-phenylethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-bromo-4-(1-methyl-1-phenylethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₂₁H₂₅BrClNO₃
MDL Number: MFCD13561328
CAS Number: 1354488-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR