Methyl (2S,4S)-4-(2-bromo-4-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050952-500MG 050952-2.5G
101893-700EA 364.62 USD
101893-700 101893-702
Methyl (2S,4S)-4-(2-bromo-4-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-bromo-4-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉BrClNO₃
MW: 364.67 g/mol
MDL Number: MFCD13561322
CAS Number: 1354488-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR