Methyl (2S,4S)-4-[2-bromo-4-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050950-500MG 050950-2.5G
101893-692EA 364.62 USD
101893-692 101893-694
Methyl (2S,4S)-4-[2-bromo-4-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-bromo-4-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₃BrClNO₃
MDL Number: MFCD13561320
CAS Number: 1354487-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR