Methyl (2S,4S)-4-[(3-bromo[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050948-500MG 050948-2.5G
101893-684EA 364.62 USD
101893-684 101893-686
Methyl (2S,4S)-4-[(3-bromo[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(3-bromo[1,1'-biphenyl]-4-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₈H₁₉BrClNO₃
MDL Number: MFCD13561318
CAS Number: 1354488-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR