(2S,4S)-4-(2-Bromo-4-isopropylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050945-500MG 050945-2.5G
101893-672EA 364.62 USD
101893-672 101893-674
(2S,4S)-4-(2-Bromo-4-isopropylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(2-Bromo-4-isopropylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₆BrNO₅
MDL Number: MFCD13561315
CAS Number: 1354485-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR