(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2,4-di(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050943-500MG 050943-2.5G
101893-664EA 364.62 USD
101893-664 101893-666
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2,4-di(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2,4-di(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₄H₃₇NO₅
MDL Number: MFCD13561313
CAS Number: 1354486-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR