(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(1,6-dibromo-2-naphthyl)oxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050935-500MG 050935-2.5G
101893-632EA 364.62 USD
101893-632 101893-634
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(1,6-dibromo-2-naphthyl)oxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(1,6-dibromo-2-naphthyl)oxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₁Br₂NO₅
MDL Number: MFCD13561305
CAS Number: 1354486-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR