Methyl (2S,4S)-4-(4-bromo-2-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050932-500MG 050932-2.5G
101893-620EA 364.62 USD
101893-620 101893-622
Methyl (2S,4S)-4-(4-bromo-2-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-bromo-2-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₁BrClNO₃
MDL Number: MFCD13561302
CAS Number: 1354488-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR