Methyl (2S,4S)-4-(2-bromo-4-chloro-3,5-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050930-500MG 050930-2.5G
101893-612EA 364.62 USD
101893-612 101893-614
Methyl (2S,4S)-4-(2-bromo-4-chloro-3,5-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-bromo-4-chloro-3,5-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈BrCl₂NO₃
MDL Number: MFCD13561300
CAS Number: 1354484-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR