Methyl (2S,4S)-4-(4-benzyl-2-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050928-500MG 050928-2.5G
101893-604EA 364.62 USD
101893-604 101893-606
Methyl (2S,4S)-4-(4-benzyl-2-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-benzyl-2-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₁BrClNO₃
MDL Number: MFCD13561298
CAS Number: 1354487-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR