(2S,4S)-4-(4-Benzyl-2-bromophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050927-500MG 050927-2.5G
101893-600EA 364.62 USD
101893-600 101893-602
(2S,4S)-4-(4-Benzyl-2-bromophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(4-Benzyl-2-bromophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₃H₂₆BrNO₅
MW: 476.38 g/mol
MDL Number: MFCD13561297
CAS Number: 1354486-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR