Methyl (2S,4S)-4-(2-bromo-4-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050924-500MG 050924-2.5G
101893-588EA 364.62 USD
101893-588 101893-590
Methyl (2S,4S)-4-(2-bromo-4-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-bromo-4-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇BrClNO₄
MDL Number: MFCD13561294
CAS Number: 1354485-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR