(2S,4S)-4-(2-Bromo-4-methoxyphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050923-500MG 050923-2.5G
101893-584EA 364.62 USD
101893-584 101893-586
(2S,4S)-4-(2-Bromo-4-methoxyphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(2-Bromo-4-methoxyphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₂BrNO₆
MDL Number: MFCD13561293
CAS Number: 1354484-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR