Methyl-3-(3-pyrrolidinyloxy)propanoate hydrochloride
Supplier: Matrix Scientific
050922-500MG
050922-2.5G
101893-580EA
364.62
USD
101893-580
101893-582
Methyl-3-(3-pyrrolidinyloxy)propanoate hydrochloride
Methyl-3-(3-pyrrolidinyloxy)propanoate hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClNO₃ |
MDL Number:
MFCD13561292 CAS Number: 1220037-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...