Methyl-3-(4-piperidinyloxy)propanoate hydrochloride

Supplier: Matrix Scientific

050919-500MG 050919-2.5G
101893-568EA 364.62 USD
101893-568 101893-570
Methyl-3-(4-piperidinyloxy)propanoate hydrochloride
Methyl-3-(4-piperidinyloxy)propanoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₃
MDL Number: MFCD13561289
CAS Number: 190515-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR