Methyl-3-(3-piperidinyloxy)propanoate hydrochloride

Supplier: Matrix Scientific

050918-500MG 050918-2.5G
101893-564EA 364.62 USD
101893-564 101893-566
Methyl-3-(3-piperidinyloxy)propanoate hydrochloride
Methyl-3-(3-piperidinyloxy)propanoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₃
MDL Number: MFCD13561288
CAS Number: 1219979-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR