3-(Cyclobutylmethoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050917-500MG 050917-2.5G
101893-560EA 364.62 USD
101893-560 101893-562
3-(Cyclobutylmethoxy)pyrrolidine hydrochloride
3-(Cyclobutylmethoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO
MDL Number: MFCD13561287
CAS Number: 1220020-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR