Cyclobutylmethyl-4-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050916-500MG 050916-2.5G
101893-556EA 364.62 USD
101893-556 101893-558
Cyclobutylmethyl-4-piperidinylmethyl ether hydrochloride
Cyclobutylmethyl-4-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClNO
MDL Number: MFCD13561286
CAS Number: 1219979-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR