4-(Cyclobutylmethoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050914-500MG 050914-2.5G
101893-548EA 364.62 USD
101893-548 101893-550
4-(Cyclobutylmethoxy)piperidine hydrochloride
4-(Cyclobutylmethoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO
MDL Number: MFCD13561284
CAS Number: 1220034-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR