3-(Cyclobutylmethoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050913-500MG 050913-2.5G
101893-544EA 364.62 USD
101893-544 101893-546
3-(Cyclobutylmethoxy)piperidine hydrochloride
3-(Cyclobutylmethoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO
MDL Number: MFCD13561283
CAS Number: 1220020-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR