4-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}piperidine hydrochloride
Supplier: Matrix Scientific
050906-500MG
050906-2.5G
101893-520EA
364.62
USD
101893-520
101893-522
4-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}piperidine hydrochloride
4-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₆ClF₃N₂O₃ |
MDL Number:
MFCD13561277 CAS Number: 1219957-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...