3-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050905-500MG 050905-2.5G
101893-516EA 364.62 USD
101893-516 101893-518
3-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}piperidine hydrochloride
3-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆ClF₃N₂O₃
MDL Number: MFCD13561276
CAS Number: 1220037-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR