3-[2-Nitro-4-(trifluoromethyl)phenoxy]piperidine hydrochloride
Supplier: Matrix Scientific
050903-500MG
050903-2.5G
101893-508EA
364.62
USD
101893-508
101893-510
3-[2-Nitro-4-(trifluoromethyl)phenoxy]piperidine hydrochloride
3-[2-Nitro-4-(trifluoromethyl)phenoxy]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₄ClF₃N₂O₃ |
MDL Number:
MFCD13561274 CAS Number: 1219957-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...