3-(2-Chloro-4-nitrophenoxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
050902-500MG
050902-2.5G
101893-504EA
364.62
USD
101893-504
101893-506
3-(2-Chloro-4-nitrophenoxy)pyrrolidine hydrochloride
3-(2-Chloro-4-nitrophenoxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₂Cl₂N₂O₃ |
MDL Number:
MFCD13561273 CAS Number: 1219979-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...