4-[(2-Chloro-4-nitrophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050901-500MG 050901-2.5G
101893-500EA 364.62 USD
101893-500 101893-502
4-[(2-Chloro-4-nitrophenoxy)methyl]piperidine hydrochloride
4-[(2-Chloro-4-nitrophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆Cl₂N₂O₃
MDL Number: MFCD13561272
CAS Number: 1220016-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR