3-[(2-Chloro-4-nitrophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050900-500MG 050900-2.5G
101893-496EA 364.62 USD
101893-496 101893-498
3-[(2-Chloro-4-nitrophenoxy)methyl]piperidine hydrochloride
3-[(2-Chloro-4-nitrophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆Cl₂N₂O₃
MDL Number: MFCD13561271
CAS Number: 1219979-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR