4-(2-Chloro-4-nitrophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050899-500MG 050899-2.5G
101893-492EA 364.62 USD
101893-492 101893-494
4-(2-Chloro-4-nitrophenoxy)piperidine hydrochloride
4-(2-Chloro-4-nitrophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₂N₂O₃
MDL Number: MFCD13561270
CAS Number: 1220037-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR