3-(2-Chloro-4-nitrophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050898-500MG 050898-2.5G
101893-488EA 364.62 USD
101893-488 101893-490
3-(2-Chloro-4-nitrophenoxy)piperidine hydrochloride
3-(2-Chloro-4-nitrophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₂N₂O₃
MDL Number: MFCD13561269
CAS Number: 1219957-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR