2-(4-Piperidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride

Supplier: Matrix Scientific

050894-500MG 050894-2.5G
101893-472EA 461.54 USD
101893-472 101893-474
2-(4-Piperidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride
2-(4-Piperidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClF₃N₂O
MDL Number: MFCD13561265
CAS Number: 1219979-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR