2-(3-Piperidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride

Supplier: Matrix Scientific

050893-500MG 050893-2.5G
101893-468EA 364.62 USD
101893-468 101893-470
2-(3-Piperidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride
2-(3-Piperidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClF₃N₂O
MDL Number: MFCD13561264
CAS Number: 1220037-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR