6-Bromo-2-pyridinyl-3-pyrrolidinyl ether hydrochloride
Supplier: Matrix Scientific
050892-500MG
050892-2.5G
101893-464EA
364.62
USD
101893-464
101893-466
6-Bromo-2-pyridinyl-3-pyrrolidinyl ether hydrochloride
6-Bromo-2-pyridinyl-3-pyrrolidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₉H₁₂BrClN₂O MW: 279.57 g/mol |
MDL Number:
MFCD13561263 CAS Number: 1220016-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...