2-Bromo-6-(3-piperidinylmethoxy)pyridine hydrochloride

Supplier: Matrix Scientific

050890-500MG 050890-2.5G
101893-456EA 364.62 USD
101893-456 101893-458
2-Bromo-6-(3-piperidinylmethoxy)pyridine hydrochloride
2-Bromo-6-(3-piperidinylmethoxy)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆BrClN₂O
MDL Number: MFCD10698906
CAS Number: 1185308-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR