2-Bromo-6-(4-piperidinyloxy)pyridine hydrochloride

Supplier: Matrix Scientific

050889-500MG 050889-2.5G
101893-452EA 364.62 USD
101893-452 101893-454
2-Bromo-6-(4-piperidinyloxy)pyridine hydrochloride
2-Bromo-6-(4-piperidinyloxy)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄BrClN₂O
MDL Number: MFCD10698902
CAS Number: 1159815-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR