2-Bromo-6-(3-piperidinyloxy)pyridine hydrochloride

Supplier: Matrix Scientific

050888-500MG 050888-2.5G
101893-448EA 226.15 USD
101893-448 101893-450
2-Bromo-6-(3-piperidinyloxy)pyridine hydrochloride
2-Bromo-6-(3-piperidinyloxy)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄BrClN₂O
MDL Number: MFCD10698904
CAS Number: 1159816-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR