5-Nitro-2-pyridinyl 3-pyrrolidinyl ether hydrochloride
Supplier: Matrix Scientific
050887-500MG
050887-2.5G
101893-444EA
364.62
USD
101893-444
101893-446
5-Nitro-2-pyridinyl 3-pyrrolidinyl ether hydrochloride
5-Nitro-2-pyridinyl 3-pyrrolidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₉H₁₂ClN₃O₃ |
MDL Number:
MFCD13561261 CAS Number: 1220020-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...