5-Nitro-2-(3-piperidinylmethoxy)pyridine hydrochloride

Supplier: Matrix Scientific

050885-500MG 050885-2.5G
101893-436EA 364.62 USD
101893-436 101893-438
5-Nitro-2-(3-piperidinylmethoxy)pyridine hydrochloride
5-Nitro-2-(3-piperidinylmethoxy)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O₃
MDL Number: MFCD09607581
CAS Number: 1185307-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR