5-Nitro-2-(4-piperidinyloxy)pyridine hydrochloride

Supplier: Matrix Scientific

050884-500MG 050884-2.5G
101893-432EA 364.62 USD
101893-432 101893-434
5-Nitro-2-(4-piperidinyloxy)pyridine hydrochloride
5-Nitro-2-(4-piperidinyloxy)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃O₃
MDL Number: MFCD09607571
CAS Number: 1185309-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR