2-Chloro-6-(3-pyrrolidinyloxy)pyrazine hydrochloride
Supplier: Matrix Scientific
050882-500MG
050882-2.5G
101893-424EA
364.62
USD
101893-424
101893-426
2-Chloro-6-(3-pyrrolidinyloxy)pyrazine hydrochloride
2-Chloro-6-(3-pyrrolidinyloxy)pyrazine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₁Cl₂N₃O |
MDL Number:
MFCD13561259 CAS Number: 1219977-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...