2-Chloro-6-(4-piperidinylmethoxy)pyrazine hydrochloride
Supplier: Matrix Scientific
050881-500MG
050881-2.5G
101893-420EA
364.62
USD
101893-420
101893-422
2-Chloro-6-(4-piperidinylmethoxy)pyrazine hydrochloride
2-Chloro-6-(4-piperidinylmethoxy)pyrazine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅Cl₂N₃O |
MDL Number:
MFCD13561258 CAS Number: 1220034-42-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...