2-Chloro-6-(3-piperidinylmethoxy)pyrazine hydrochloride
Supplier: Matrix Scientific
050880-500MG
050880-2.5G
101893-416EA
364.62
USD
101893-416
101893-418
2-Chloro-6-(3-piperidinylmethoxy)pyrazine hydrochloride
2-Chloro-6-(3-piperidinylmethoxy)pyrazine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅Cl₂N₃O |
MDL Number:
MFCD13561257 CAS Number: 1220036-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...