6-Chloro-2-pyrazinyl-4-piperidinyl ether hydrochloride
Supplier: Matrix Scientific
050879-500MG
050879-2.5G
101893-412EA
364.62
USD
101893-412
101893-414
6-Chloro-2-pyrazinyl-4-piperidinyl ether hydrochloride
6-Chloro-2-pyrazinyl-4-piperidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃Cl₂N₃O MW: 250.13 g/mol |
MDL Number:
MFCD13561256 CAS Number: 1220036-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...