6-Chloro-2-pyrazinyl-3-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050878-500MG 050878-2.5G
101893-408EA 364.62 USD
101893-408 101893-410
6-Chloro-2-pyrazinyl-3-piperidinyl ether hydrochloride
6-Chloro-2-pyrazinyl-3-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₉H₁₃Cl₂N₃O
MW: 250.13 g/mol
MDL Number: MFCD13561255
CAS Number: 1220020-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR