6-Chloro-4-pyrimidinyl-4-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050874-500MG 050874-2.5G
101893-392EA 364.62 USD
101893-392 101893-394
6-Chloro-4-pyrimidinyl-4-piperidinyl ether hydrochloride
6-Chloro-4-pyrimidinyl-4-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₉H₁₃Cl₂N₃O
MW: 250.13 g/mol
MDL Number: MFCD09607626
CAS Number: 1185314-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR